2-Chloro-N-[4-(2-chlorophenyl)-1,3-thiazol-2-yl]acetamide
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:338957-50-9
:
Formula:C₁₁H₈Cl₂N₂OS
MDL Number:MFCD01315353
:287.17 g/mol
Melting Pt:130…132 °C
Storage Temperature:Ambient
Frequently Bought Together