
2'-Chloro-4'-(4-chlorophenoxy)acetophenone 98+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:119851-28-4
:
Formula:C₁₄H₁₀Cl₂O₂
MDL Number:MFCD00140226
Melting Pt:54…58 °C
:281.1340 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together