Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(2-Amino-5-morpholinophenyl)-1-ethanone
1-(2-Amino-5-morpholinophenyl)-1-ethanone 95%
1-(2-Amino-5-morpholinophenyl)-1-ethanone 95%
APOSOR15306-500MG
:
Apollo Scientific
:
1-(2-Amino-5-morpholinophenyl)-1-ethanone 95%
APOSOR15306-500MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
500 mg
Seq:
2725
:
:
220.27 g/mol
:
104…105 °C
:
GHS07
:
Ambient
:
98440-50-7
:
C₁₂H₁₆N₂O₂
:
MFCD03787743