
1-(5-Chloro-2-nitrophenyl)-1-ethanone
Catalog # APOSOR15281-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:199.59 g/mol
Melting Pt:58…61 °C
:GHS07
Storage Temperature:Ambient
CAS Number:18640-60-3
Formula:C₈H₆ClNO₃
MDL Number:MFCD03787322