Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Boc-4-(2-chloroethyl)piperazine
1-Boc-4-(2-chloroethyl)piperazine
1-Boc-4-(2-chloroethyl)piperazine
APOSOR15245-1G
:
Apollo Scientific
:
1-Boc-4-(2-chloroethyl)piperazine
APOSOR15245-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
248.751 g/mol
:
63…65 °C
:
GHS07
:
Ambient
:
208167-83-3
:
C₁₁H₂₁ClN₂O₂
:
MFCD08443993