
Methyl-4-(2-oxo-1-imidazolidinyl)benzenecarboxylate
Catalog # APOSOR15226-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:220.23 g/mol
Melting Pt:219…221 °C
:GHS07
Storage Temperature:Ambient
CAS Number:627901-54-6
Formula:C₁₁H₁₂N₂O₃
MDL Number:MFCD08443989