
5-Amino-3-(4-methoxyphenyl)-1H-pyrazole
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:19541-95-8
:
Formula:C₁₀H₁₁N₃O
MDL Number:MFCD00462192
:189.22 g/mol
Melting Pt:140…144 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together