
3-(3-Methoxyphenyl)-3-oxopropanenitrile 98%
Catalog # APOSOR15211-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:175.19 g/mol
Melting Pt:89…91 °C
:GHS07
Storage Temperature:Ambient
CAS Number:21667-60-7
Formula:C₁₀H₉NO₂
MDL Number:MFCD01566517