Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl-2-(1H-pyrazol-1-yl)acetate
Ethyl-2-(1H-pyrazol-1-yl)acetate
Ethyl-2-(1H-pyrazol-1-yl)acetate
Ethyl-2-(1H-pyrazol-1-yl)acetate
APOSOR15197-250MG
:
Apollo Scientific
:
Ethyl-2-(1H-pyrazol-1-yl)acetate
APOSOR15197-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
154.17 g/mol
:
GHS07
:
Ambient
:
10199-61-8
:
C₇H₁₀N₂O₂
:
MFCD00159620