
5,7-Dimethyl[1,2,4]triazolo[1,5-a]pyrimidin-2-ylamine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:7135-02-6
:
Formula:C₇H₉N₅
MDL Number:MFCD00227025
Melting Pt:360…363 °C
:163.18 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together