Order Entry
Czech Republic
ContactUsLinkComponent
 
Irritant
3-(3,4-Dimethoxyphenyl)-1H-pyrazol-5-amine
Catalog # APOSOR15171-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-(3,4-Dimethoxyphenyl)-1H-pyrazol-5-amine
Catalog # APOSOR15171-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Melting Pt:131…133 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:208519-08-8
Formula:C₁₁H₁₃N₃O₂
MDL Number:MFCD02664228