4-Chloro-2-[(dimethylamino)methyl]-6-methylbenzenol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:87117-40-6
:
Formula:C₁₀H₁₄ClNO
MDL Number:MFCD08443991
:199.68 g/mol
Storage Temperature:Ambient
Frequently Bought Together