
5-Nitro-1,2-dihydro-3H-indazol-3-one
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:61346-19-8
:
Formula:C₇H₅N₃O₃
MDL Number:MFCD00012418
:179.14 g/mol
Melting Pt:280…282 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together