Methyl-4-(3-nitro-2-pyridinyl)benzenecarboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:937601-36-0
:
Formula:C₁₃H₁₀N₂O₄
MDL Number:MFCD08443945
Melting Pt:82…84 °C
:258.24 g/mol
Storage Temperature:Ambient
Frequently Bought Together