Order Entry
Czech Republic
ContactUsLinkComponent
Ethyl-4-hydroxy-2-(4-methoxyphenyl)-1,3-thiazole-5-carboxylate
Irritant
Ethyl-4-hydroxy-2-(4-methoxyphenyl)-1,3-thiazole-5-carboxylate
Catalog # APOSOR15092-500MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Ethyl-4-hydroxy-2-(4-methoxyphenyl)-1,3-thiazole-5-carboxylate
Catalog # APOSOR15092-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :279.32 g/mol
Melting Pt:147…149 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:924869-01-2
Formula:C₁₃H₁₃NO₄S
MDL Number:MFCD08444006