
Ethyl-2-(3,4-dimethoxyphenyl)-4-hydroxy-1,3-thiazole-5-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:924868-99-5
:
Formula:C₁₄H₁₅NO₅S
MDL Number:MFCD08444004
Melting Pt:144…146 °C
:309.34 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together