1-(1-Acetyl-1H-indol-3-yl)-1-ethanone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:17537-64-3
:
Formula:C₁₂H₁₁NO₂
MDL Number:MFCD00039692
:201.23 g/mol
Melting Pt:153…156 °C
Storage Temperature:Ambient
Frequently Bought Together