Order Entry
Czech Republic
ContactUsLinkComponent
Methyl-2-methyl-1,3-benzoxazole-5-carboxylate
Irritant
Methyl-2-methyl-1,3-benzoxazole-5-carboxylate
Catalog # APOSOR15047-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Methyl-2-methyl-1,3-benzoxazole-5-carboxylate
Catalog # APOSOR15047-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :191.19 g/mol
Melting Pt:66…68 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:136663-21-3
Formula:C₁₀H₉NO₃
MDL Number:MFCD07774245