Order Entry
Czech Republic
ContactUsLinkComponent
1-(4-Cyanophenyl)-2-thiourea 97%
Irritant
1-(4-Cyanophenyl)-2-thiourea 97%
Catalog # APOSOR15046-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-(4-Cyanophenyl)-2-thiourea 97%
Catalog # APOSOR15046-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :177.23 g/mol
Melting Pt:201…205 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:3460-55-7
Formula:C₈H₇N₃S
MDL Number:MFCD00041175