

2-Tetrahydro-2H-pyran-4-ylethanol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:4677-18-3
:
Formula:C₇H₁₄O₂
MDL Number:MFCD00129068
:130.19 g/mol
Boiling Pt:119…120 °C/14mmHg
UN:1987
ADR:3,III
:GHS02
:GHS07
Storage Temperature:Ambient
Frequently Bought Together