
4-(3,5-Dimethyl-1H-pyrazol-1-yl)benzonitrile 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:56935-79-6
:
Formula:C₁₂H₁₁N₃
MDL Number:MFCD09817559
Melting Pt:57…59 °C
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
:GHS07
Frequently Bought Together