Order Entry
Czech Republic
ContactUsLinkComponent
1-(4-Bromophenyl)-3,5-dimethyl-1H-pyrazole 97%
Irritant
1-(4-Bromophenyl)-3,5-dimethyl-1H-pyrazole 97%
Catalog # APOSOR14885-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-(4-Bromophenyl)-3,5-dimethyl-1H-pyrazole 97%
Catalog # APOSOR14885-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :251.13 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:62546-27-4
Formula:C₁₁H₁₁BrN₂
MDL Number:MFCD09817558