
Tetrahydro-2H-pyran-4-ylacetyl chloride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:40500-05-8
:
Formula:C₇H₁₁ClO₂
MDL Number:MFCD08669649
:162.62 g/mol
Boiling Pt:110…111 °C/11mmHg
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05
Frequently Bought Together