0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
2-[(4-Chlorobenzyl)amino]benzenecarboxylic acid
2-[(4-Chlorobenzyl)amino]benzenecarboxylic acid
2-[(4-Chlorobenzyl)amino]benzenecarboxylic acid
APOSOR14814-1G
:
Apollo Scientific
:
2-[(4-Chlorobenzyl)amino]benzenecarboxylic acid
APOSOR14814-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
261.71 g/mol
:
146 °C
:
GHS07
:
Ambient
:
14345-04-1
:
C₁₄H₁₂ClNO₂
:
MFCD03032901