

2-Bromo-4-chlorobenzoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:936-08-3
:
Formula:C₇H₄BrClO₂
MDL Number:MFCD00672930
:235.46 g/mol
Melting Pt:157…161 °C
UN:2811
ADR:6.1,III
:GHS07
:GHS09
Storage Temperature:Ambient
Frequently Bought Together