
3-(2-Pyridyl)-L-alanine 99+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:37535-51-6
:
Formula:C₈H₁₀N₂O₂
MDL Number:MFCD00237827
:166.18 g/mol
Melting Pt:191.4 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together