2-[(4,6-Dimethoxypyrimidin-2-yl)thio]benzoic acid 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:110284-79-2
:
Formula:C₁₃H₁₂N₂O₄S
MDL Number:MFCD00203074
:292.32 g/mol
Melting Pt:129…130 °C
Storage Temperature:Ambient
Frequently Bought Together