
Ethyl pyrazolo[1,5-a]pyridine-3-carboxylate 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:16205-44-0
:
Formula:C₁₀H₁₀N₂O₂
MDL Number:MFCD06411542
:190.1986 g/mol
Boiling Pt:134…135 °C/2.3mmHg
Density:1.23 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together