
2-Chloro-1-(3,4-dibromo-2-thienyl)-1-ethanone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:62673-71-6
:
Formula:C₆H₃Br₂ClOS
MDL Number:MFCD06411631
:318.43 g/mol
Melting Pt:96…98 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together