
3-(1,3-Dioxan-2-yl)aniline 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:168551-56-2
:
Formula:C₁₀H₁₃NO₂
MDL Number:MFCD02853017
:179.22 g/mol
Melting Pt:75…76 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together