
5-(Chloromethyl)-2-(4-methoxyphenyl)pyridine
Catalog # APOSOR14197-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:233.7 g/mol
Melting Pt:46…49 °C
:GHS07
Storage Temperature:Ambient
CAS Number:886361-72-4
Formula:C₁₃H₁₂ClNO
MDL Number:MFCD06200955