2-(Chloromethyl)-1-(4-chlorophenoxy)-4-nitrobenzene
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:924862-19-1
:
Formula:C₁₃H₉Cl₂NO₃
MDL Number:MFCD08689723
:298.13 g/mol
Melting Pt:90…92 °C
Storage Temperature:Ambient
Frequently Bought Together