Order Entry
Czech Republic
ContactUsLinkComponent
2-Bromo-1-(5-chloro-2-thienyl)-1-ethanone
Corrosive
Irritant
2-Bromo-1-(5-chloro-2-thienyl)-1-ethanone
Catalog # APOSOR14033-1G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
2-Bromo-1-(5-chloro-2-thienyl)-1-ethanone
Catalog # APOSOR14033-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :239.52 g/mol
Melting Pt:73…75 °C
Storage Temperature:Refrigerator
UN:1759
ADR:8,III
 :GHS05
 :GHS07
CAS Number:57731-17-6
Formula:C₆H₄BrClOS
MDL Number:MFCD00829820