Order Entry
Czech Republic
ContactUsLinkComponent
5-Iodo-2-(4-morpholino)pyridine
Irritant
5-Iodo-2-(4-morpholino)pyridine
Catalog # APOSOR14007-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-Iodo-2-(4-morpholino)pyridine
Catalog # APOSOR14007-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :290.11 g/mol
Melting Pt:129…135 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:470463-42-4
Formula:C₉H₁₁IN₂O
MDL Number:MFCD04039866