
1,2-Dibromocyclopentene, Technical Grade
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:75415-78-0
:
Formula:C₅H₆Br₂
MDL Number:MFCD00010246
:225.911 g/mol
Boiling Pt:78 °C/5mmHg
Density:1.895 g/cm³ (25 °C)
Flash Pt:>113 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together