
1-(2,4-Dichlorophenyl)-2-(1-imidazolyl)ethanol
Catalog # APOSOR13643-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:257.11 g/mol
Melting Pt:135 °C
EINECS:246-042-5
:GHS07
Storage Temperature:Ambient
CAS Number:24155-42-8
Formula:C₁₁H₁₀Cl₂N₂O
MDL Number:MFCD00044708