

2,6-Dimethylpiperidin (mixture of cis- and trans-isomers)
Catalog # APOSOR13600-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:113.2 g/mol
Boiling Pt:126…129 °C
Melting Pt:< –20 °C
Density:0.832 g/cm³ (25 °C)
Flash Pt:11 °C
Storage Temperature:Ambient
EINECS:207-981-6
UN:2733
ADR:3+8,II
:GHS02
:GHS05
CAS Number:504-03-0
Formula:C₇H₁₅N
MDL Number:MFCD00066304