
3-Nitro-1H-pyrazole-4-carbonitrile
Catalog # APOSOR13556-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:138.0800 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:39205-87-3
Formula:C4H2N4O2
MDL Number:MFCD00195039