
Methyl-4-chloropicolinate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:24484-93-3
:
Formula:C₇H₆ClNO₂
MDL Number:MFCD04116183
:171.5800 g/mol
Melting Pt:56…58 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together