
Methyl-2-amino-5-chlorobenzoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:5202-89-1
:
Formula:C₈H₈ClNO₂
MDL Number:MFCD00007837
:185.61 g/mol
Boiling Pt:168…170 °C/22mmHg
Melting Pt:66…70 °C
Storage Temperature:Ambient
EINECS:225-992-4
:GHS07
Frequently Bought Together