
{2-[(4-Chlorophenyl)sulphonyl]ethyl}amine hydrochloride
Catalog # APOSOR12933-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:256.15 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:85052-88-6
Formula:C₈H₁₁Cl₂NO₂S
MDL Number:MFCD03840166