
2-Nitro-1,3,5-trimethoxybenzene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:14227-18-0
:
Formula:C₉H₁₁NO₅
MDL Number:MFCD00016992
:213.19 g/mol
Melting Pt:149…151 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together