
3-(2-Pyrrolidinyl)pyridine
Catalog # APOSOR12516-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:148.21 g/mol
Boiling Pt:107…108 °C/2mmHg
Density:1.074 g/cm³ (25 °C)
Flash Pt:101 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:5746-86-1
Formula:C₉H₁₂N₂
MDL Number:MFCD00016913