Order Entry
Czech Republic
ContactUsLinkComponent
[(5-Methyl-2-thienyl)methyl]amine hydrochloride 97%
Toxic
[(5-Methyl-2-thienyl)methyl]amine hydrochloride 97%
Catalog # APOSOR12377-100MG
Supplier:  Apollo Scientific
CAS Number:  
Toxic
[(5-Methyl-2-thienyl)methyl]amine hydrochloride 97%
Catalog # APOSOR12377-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
Melting Pt:191…193 °C
 :163.67 g/mol
 :GHS06
Storage Temperature:Ambient
CAS Number:171661-55-5
Formula:C₆H₁₀ClNS
MDL Number:MFCD09817548