Order Entry
Czech Republic
ContactUsLinkComponent
4-(1-Methyl-1H-pyrazol-3-yl)benzonitrile 97%
Irritant
4-(1-Methyl-1H-pyrazol-3-yl)benzonitrile 97%
Catalog # APOSOR12364-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-(1-Methyl-1H-pyrazol-3-yl)benzonitrile 97%
Catalog # APOSOR12364-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
Melting Pt:146…147 °C
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
 :GHS07
CAS Number:915707-41-4
Formula:C₁₁H₉N₃
MDL Number:MFCD09702350