Order Entry
Czech Republic
ContactUsLinkComponent
3-Furylmethylamine hydrochloride 97%
Irritant
3-Furylmethylamine hydrochloride 97%
Catalog # APOSOR12311-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-Furylmethylamine hydrochloride 97%
Catalog # APOSOR12311-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :133.58 g/mol
Melting Pt:167…168 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:131052-43-2
Formula:C₅H₈ClNO
MDL Number:MFCD06407993