
2-(Tetrahydro-2H-pyran-4-yloxy)pyridine-5-boronic acid pinacol ester
Catalog # APOSOR12267-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:305.18 g/mol
Melting Pt:112…113.5 °C
:GHS07
Storage Temperature:Ambient
CAS Number:910036-98-5
Formula:C₁₆H₂₄BNO₄
MDL Number:MFCD09702390