
4-(1-Methyl-1H-pyrazol-3-yl)phenylamine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:916766-82-0
:
Formula:C₁₀H₁₁N₃
MDL Number:MFCD03407387
:173.22 g/mol
Melting Pt:125…133 °C
Storage Temperature:Refrigerator
UN:2811
ADR:6.1,III
:GHS07
Frequently Bought Together