
2-(2-Methylimidazol-1-yl)phenylamine 95+%
Catalog # APOSOR12232-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:173.22 g/mol
Melting Pt:132.5…134.5 °C
:GHS07
Storage Temperature:Ambient
CAS Number:26286-55-5
Formula:C₁₀H₁₁N₃
MDL Number:MFCD08699292