
Methyl-3-(2-bromoethoxy)benzenecarboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:59516-96-0
:
Formula:C₁₀H₁₁BrO₃
MDL Number:MFCD06204041
Boiling Pt:110 °C/0.038mmHg
:259.11 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together